Assay Details
| Gene Symbol: ppm1k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327744.1 | NP_001314673.1 | 6 - 7 | 1198 | 66 | |
| XM_005160091.4 | XP_005160148.1 | 6 - 7 | 1219 | 66 | ||
| XM_005160093.4 | XP_005160150.1 | 4 - 5 | 1161 | 66 | ||
| GenBank mRNA | BC090752.1 | - | 6 - 7 | 590 | 66 | |
More Information
Set Membership: |
|

ppm1k
Genomic Map