Assay Details
| Gene Symbol: eps8l1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013313.1 | NP_001013331.1 | 11 - 12 | 1186 | 75 | |
| XM_009299281.3 | XP_009297556.1 | 12 - 13 | 1157 | 75 | ||
| XM_009299282.3 | XP_009297557.1 | 11 - 12 | 1113 | 75 | ||
| XM_009299283.3 | XP_009297558.1 | 11 - 12 | 1233 | 75 | ||
| XM_021470513.1 | XP_021326188.1 | 10 - 11 | 939 | 75 | ||
| GenBank mRNA | BC090754.1 | - | 11 - 12 | 1186 | 75 | |
| BC154166.1 | - | 11 - 12 | 1171 | 75 | ||
More Information
Set Membership: |
|

eps8l1a
Genomic Map