Assay Details
| Gene Symbol: tmem182a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013320.1 | NP_001013338.1 | 4 - 5 | 740 | 70 | |
| GenBank mRNA | BC090696.1 | - | 4 - 5 | 740 | 70 | |
More Information
Set Membership: |
|

tmem182a
Genomic Map