Assay Details
| Gene Symbol: ptrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013330.2 | NP_001013348.1 | 4 - 5 | 666 | 68 | |
| GenBank mRNA | BC090788.1 | - | 4 - 5 | 670 | 68 | |
| BC165428.1 | - | 3 - 4 | 420 | 68 | ||
More Information
Set Membership: |
|

ptrh1
Genomic Map