Assay Details
| Gene Symbol: ptrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013330.2 | NP_001013348.1 | 5 - 6 | 711 | 103 | |
| GenBank mRNA | BC090788.1 | - | 5 - 6 | 715 | 103 | |
| BC165428.1 | - | 4 - 5 | 465 | 103 | ||
More Information
Set Membership: |
|

ptrh1
Genomic Map