Assay Details
| Gene Symbol: msi1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013333.1 | NP_001013351.1 | 2 - 3 | 133 | 75 | |
| XM_005171742.4 | XP_005171799.1 | 2 - 3 | 368 | 75 | ||
| GenBank mRNA | BC071308.1 | - | 2 - 3 | 130 | 75 | |
| BC090772.1 | - | 2 - 3 | 133 | 75 | ||
| Gene Symbol: LOC100537556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003198578.5 | XP_003198626.3 | 214 | 75 | ||
More Information
Set Membership: |
|

msi1a
Genomic Map