Assay Details
| Gene Symbol: tp53i11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013352.2 | NP_001013370.1 | 7 - 8 | 641 | 76 | |
| GenBank mRNA | BC090705.1 | - | 7 - 8 | 645 | 76 | |
More Information
Set Membership: |
|

tp53i11a
Genomic Map