Assay Details
| Gene Symbol: itga6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013448.1 | NP_001013466.1 | 24 - 25 | 3146 | 73 | |
| XM_009291986.3 | XP_009290261.1 | 24 - 25 | 167 | |||
| GenBank mRNA | BC090458.1 | - | 24 - 25 | 3146 | 73 | |
More Information
Set Membership: |
|

itga6b
Genomic Map