Assay Details
| Gene Symbol: ndufa9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013459.2 | NP_001013477.2 | 9 - 10 | 964 | 78 | |
| GenBank mRNA | BC068378.1 | - | 9 - 10 | 892 | 78 | |
| BC091545.1 | - | 9 - 10 | 913 | 78 | ||
| BC152139.1 | - | 9 - 10 | 914 | 78 | ||
| BC153422.1 | - | 9 - 10 | 1364 | 78 | ||
More Information
Set Membership: |
|

ndufa9a
Genomic Map