Assay Details
| Gene Symbol: capn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013501.2 | NP_001013519.1 | 19 - 20 | 2099 | 63 | |
| GenBank mRNA | BC090435.1 | - | 19 - 20 | 2103 | 63 | |
More Information
Set Membership: |
|

capn2a
Genomic Map