Assay Details
| Gene Symbol: mybpc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013511.2 | NP_001013529.2 | 6 - 7 | 437 | 70 | |
| XM_017353828.2 | XP_017209317.1 | 5 - 6 | 502 | 70 | ||
| XM_021470159.1 | XP_021325834.1 | 6 - 7 | 471 | 70 | ||
| XM_021470160.1 | XP_021325835.1 | 5 - 6 | 499 | 70 | ||
| XM_021470161.1 | XP_021325836.1 | 5 - 6 | 449 | 70 | ||
| XM_021470162.1 | XP_021325837.1 | 5 - 6 | 386 | 70 | ||
| GenBank mRNA | BC091662.1 | - | 431 | 70 | ||
| CK676323.1 | - | 6 - 7 | 431 | 70 | ||
| EB790963.1 | - | 2 - 3 | 204 | 70 | ||
More Information
Set Membership: |
|

mybpc2b
Genomic Map