Assay Details
| Gene Symbol: mybpc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013511.2 | NP_001013529.2 | 18 - 19 | 1842 | 97 | |
| XM_017353828.2 | XP_017209317.1 | 17 - 18 | 1907 | 97 | ||
| XM_021470159.1 | XP_021325834.1 | 18 - 19 | 1876 | 97 | ||
| XM_021470160.1 | XP_021325835.1 | 17 - 18 | 1904 | 97 | ||
| XM_021470161.1 | XP_021325836.1 | 17 - 18 | 1854 | 97 | ||
| XM_021470162.1 | XP_021325837.1 | 17 - 18 | 1791 | 97 | ||
| GenBank mRNA | BC091662.1 | - | 97 | |||
| EE714901.1 | - | 2 - 3 | 200 | 97 | ||
More Information
Set Membership: |
|

mybpc2b
Genomic Map