Assay Details
| Gene Symbol: mybpc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013511.2 | NP_001013529.2 | 22 - 23 | 2438 | 104 | |
| XM_017353828.2 | XP_017209317.1 | 21 - 22 | 2503 | 104 | ||
| XM_021470159.1 | XP_021325834.1 | 22 - 23 | 2472 | 104 | ||
| XM_021470160.1 | XP_021325835.1 | 21 - 22 | 2500 | 104 | ||
| XM_021470161.1 | XP_021325836.1 | 21 - 22 | 2450 | 104 | ||
| XM_021470162.1 | XP_021325837.1 | 21 - 22 | 2387 | 104 | ||
| GenBank mRNA | BC091662.1 | - | 2432 | 104 | ||
| EB866488.1 | - | 1 - 2 | 162 | 104 | ||
More Information
Set Membership: |
|

mybpc2b
Genomic Map