Assay Details
| Gene Symbol: mybpc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013511.2 | NP_001013529.2 | 25 - 26 | 2864 | 88 | |
| XM_017353828.2 | XP_017209317.1 | 24 - 25 | 2929 | 88 | ||
| XM_021470159.1 | XP_021325834.1 | 25 - 26 | 2898 | 88 | ||
| XM_021470160.1 | XP_021325835.1 | 24 - 25 | 2926 | 88 | ||
| XM_021470161.1 | XP_021325836.1 | 24 - 25 | 2876 | 88 | ||
| XM_021470162.1 | XP_021325837.1 | 24 - 25 | 2813 | 88 | ||
| GenBank mRNA | BC091662.1 | - | 2858 | 88 | ||
| CK677061.1 | - | 2 - 3 | 196 | 88 | ||
More Information
Set Membership: |
|

mybpc2b
Genomic Map