Assay Details
| Gene Symbol: mybpc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013511.2 | NP_001013529.2 | 4 - 5 | 161 | 85 | |
| XM_017353828.2 | XP_017209317.1 | 3 - 4 | 226 | 85 | ||
| XM_021470159.1 | XP_021325834.1 | 4 - 5 | 195 | 85 | ||
| XM_021470160.1 | XP_021325835.1 | 3 - 4 | 223 | 85 | ||
| XM_021470161.1 | XP_021325836.1 | 3 - 4 | 173 | 85 | ||
| XM_021470162.1 | XP_021325837.1 | 3 - 4 | 110 | 85 | ||
| GenBank mRNA | BC091662.1 | - | 155 | 85 | ||
| CK676323.1 | - | 4 - 5 | 155 | 85 | ||
| EB776017.1 | - | 4 - 5 | 164 | 85 | ||
More Information
Set Membership: |
|

mybpc2b
Genomic Map