Assay Details
| Gene Symbol: mybpc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013511.2 | NP_001013529.2 | 12 - 13 | 1073 | 77 | |
| XM_017353828.2 | XP_017209317.1 | 11 - 12 | 1138 | 77 | ||
| XM_021470159.1 | XP_021325834.1 | 12 - 13 | 1107 | 77 | ||
| XM_021470160.1 | XP_021325835.1 | 11 - 12 | 1135 | 77 | ||
| XM_021470161.1 | XP_021325836.1 | 11 - 12 | 1085 | 77 | ||
| XM_021470162.1 | XP_021325837.1 | 11 - 12 | 1022 | 77 | ||
| GenBank mRNA | BC091662.1 | - | 1067 | 77 | ||
| EE714531.1 | - | 4 - 5 | 501 | 77 | ||
| EE719585.1 | - | 2 - 3 | 201 | 77 | ||
More Information
Set Membership: |
|

mybpc2b
Genomic Map