Assay Details
| Gene Symbol: mat2ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014296.2 | NP_001014318.1 | 2 - 3 | 233 | 68 | |
| XM_017356170.2 | XP_017211659.1 | 2 - 3 | 246 | 68 | ||
| GenBank mRNA | BC164405.1 | - | 2 - 3 | 206 | 68 | |
| JN848835.1 | - | 2 - 3 | 229 | 68 | ||
More Information
Set Membership: |
|

mat2ab
Genomic Map