Assay Details
| Gene Symbol: mat2ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014296.2 | NP_001014318.1 | 3 - 4 | 355 | 74 | |
| XM_017356170.2 | XP_017211659.1 | 3 - 4 | 368 | 74 | ||
| GenBank mRNA | BC164405.1 | - | 3 - 4 | 328 | 74 | |
| JN848835.1 | - | 3 - 4 | 351 | 74 | ||
More Information
Set Membership: |
|

mat2ab
Genomic Map