Assay Details
| Gene Symbol: trim2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014371.1 | NP_001014393.1 | 7 - 8 | 1748 | 104 | |
| XM_005170889.4 | XP_005170946.1 | 7 - 8 | 104 | |||
| XM_005170890.4 | XP_005170947.1 | 3 - 4 | 104 | |||
| GenBank mRNA | BC091789.1 | - | 7 - 8 | 1748 | 104 | |
More Information
Set Membership: |
|

trim2a
Genomic Map