Assay Details
| Gene Symbol: trim2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014371.1 | NP_001014393.1 | 9 - 10 | 2085 | 114 | |
| XM_005170889.4 | XP_005170946.1 | 9 - 10 | 2164 | 114 | ||
| XM_005170890.4 | XP_005170947.1 | 5 - 6 | 1257 | 114 | ||
| GenBank mRNA | BC091789.1 | - | 9 - 10 | 2085 | 114 | |
More Information
Set Membership: |
|

trim2a
Genomic Map