Assay Details
| Gene Symbol: trim2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014371.1 | NP_001014393.1 | 10 - 11 | 2156 | 141 | |
| XM_005170889.4 | XP_005170946.1 | 10 - 11 | 2235 | 141 | ||
| XM_005170890.4 | XP_005170947.1 | 6 - 7 | 1328 | 141 | ||
| GenBank mRNA | BC091789.1 | - | 10 - 11 | 2156 | 141 | |
More Information
Set Membership: |
|

trim2a
Genomic Map