Assay Details
| Gene Symbol: snx18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015063.1 | NP_001015063.1 | 1 - 2 | 1644 | 75 | |
| GenBank mRNA | AY078363.1 | - | 1 - 2 | 1644 | 75 | |
| BC162101.1 | - | 1 - 2 | 1514 | 75 | ||
More Information
Set Membership: |
|

snx18b
Genomic Map