Assay Details
| Gene Symbol: tmem11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017400.1 | NP_001017400.1 | 1 - 2 | 61 | 68 | |
| GenBank mRNA | BC067587.1 | - | 1 - 2 | 48 | 68 | |
| BC092726.1 | - | 1 - 2 | 61 | 68 | ||
More Information
Set Membership: |
|

tmem11
Genomic Map