Assay Details
| Gene Symbol: actr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017542.2 | NP_001017542.1 | 6 - 7 | 823 | 74 | |
| XM_005169510.4 | XP_005169567.1 | 6 - 7 | 848 | 74 | ||
| XM_021480582.1 | XP_021336257.1 | 7 - 8 | 757 | 74 | ||
| GenBank mRNA | BC092187.1 | - | 6 - 7 | 827 | 74 | |
| BC153558.1 | - | 6 - 7 | 747 | 74 | ||
More Information
Set Membership: |
|

actr2b
Genomic Map