Assay Details
| Gene Symbol: ca8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017571.2 | NP_001017571.2 | 4 - 5 | 900 | 75 | |
| GenBank mRNA | BC092740.1 | - | 4 - 5 | 782 | 75 | |
| BC164636.1 | - | 4 - 5 | 521 | 75 | ||
More Information
Set Membership: |
|

ca8
Genomic Map