Assay Details
| Gene Symbol: ca8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017571.2 | NP_001017571.2 | 7 - 8 | 1122 | 88 | |
| GenBank mRNA | BC092740.1 | - | 7 - 8 | 1004 | 88 | |
| BC164636.1 | - | 7 - 8 | 743 | 88 | ||
More Information
Set Membership: |
|

ca8
Genomic Map