Assay Details
| Gene Symbol: cdca7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017574.1 | NP_001017574.1 | 6 - 7 | 846 | 94 | |
| XM_005167416.4 | XP_005167473.1 | 6 - 7 | 907 | 94 | ||
| GenBank mRNA | BC092737.1 | - | 6 - 7 | 846 | 94 | |
More Information
Set Membership: |
|

cdca7a
Genomic Map