Assay Details
Gene Symbol: ankrd49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017584.2 | NP_001017584.2 | 2 - 3 | 253 | 85 | |
GenBank mRNA | BC092998.1 | - | 2 - 3 | 244 | 85 | |
EH500845.1 | - | 2 - 3 | 278 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
