Assay Details
| Gene Symbol: cpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017591.1 | NP_001017591.1 | 2 - 3 | 361 | 69 | |
| GenBank mRNA | BC092863.1 | - | 2 - 3 | 361 | 69 | |
More Information
Set Membership: |
|

cpm
Genomic Map