Assay Details
Gene Symbol: lgals3bp.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017658.1 | NP_001017658.1 | 3 - 4 | 450 | 72 | |
GenBank mRNA | BC093341.1 | - | 3 - 4 | 450 | 72 | |
BC164862.1 | - | 3 - 4 | 430 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
