Assay Details
| Gene Symbol: dync1li2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017669.2 | NP_001017669.2 | 9 - 10 | 1088 | 63 | |
| XM_005166450.4 | XP_005166507.1 | 9 - 10 | 1126 | 63 | ||
| GenBank mRNA | BC093336.1 | - | 9 - 10 | 63 | ||
| BC154268.1 | - | 9 - 10 | 63 | |||
| BC165621.1 | - | 9 - 10 | 63 | |||
More Information
Set Membership: |
|

dync1li2
Genomic Map