Assay Details
| Gene Symbol: zgc:112023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017673.1 | NP_001017673.1 | 4 - 5 | 749 | 84 | |
| XM_009302157.3 | XP_009300432.1 | 4 - 5 | 827 | 84 | ||
| XM_009302158.3 | XP_009300433.1 | 3 - 4 | 693 | 84 | ||
| GenBank mRNA | BC093162.1 | - | 4 - 5 | 749 | 84 | |
More Information
Set Membership: |
|

zgc:112023
Genomic Map