Assay Details
| Gene Symbol: capsla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017676.1 | NP_001017676.1 | 2 - 3 | 112 | 57 | |
| XM_009298923.3 | XP_009297198.1 | 2 - 3 | 156 | 57 | ||
| GenBank mRNA | BC093331.1 | - | 2 - 3 | 112 | 57 | |
More Information
Set Membership: |
|

capsla
Genomic Map