Assay Details
| Gene Symbol: osr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017694.1 | NP_001017694.1 | 2 - 3 | 718 | 76 | |
| XM_005173555.4 | XP_005173612.1 | 2 - 3 | 734 | 76 | ||
| GenBank mRNA | BC093148.1 | - | 2 - 3 | 718 | 76 | |
| BC165157.1 | - | 1 - 2 | 573 | 76 | ||
More Information
Set Membership: |
|

osr2
Genomic Map