Assay Details
| Gene Symbol: mrm3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017695.2 | NP_001017695.2 | 3 - 4 | 806 | 119 | |
| GenBank mRNA | BC093318.1 | - | 3 - 4 | 798 | 119 | |
| BC164530.1 | - | 3 - 4 | 823 | 119 | ||
| EH576416.1 | - | 1 - 2 | 560 | 119 | ||
More Information
Set Membership: |
|

mrm3a
Genomic Map