Assay Details
| Gene Symbol: magoh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017700.1 | NP_001017700.1 | 4 - 5 | 411 | 123 | |
| GenBank mRNA | BC093273.1 | - | 4 - 5 | 411 | 123 | |
More Information
Set Membership: |
|

magoh
Genomic Map