Assay Details
| Gene Symbol: pdrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017757.2 | NP_001017757.1 | 2 - 3 | 187 | 93 | |
| GenBank mRNA | BC093358.1 | - | 2 - 3 | 171 | 93 | |
| EH544878.1 | - | 2 - 3 | 194 | 93 | ||
More Information
Set Membership: |
|

pdrg1
Genomic Map