Assay Details
| Gene Symbol: lrrc39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017760.1 | NP_001017760.1 | 3 - 4 | 495 | 96 | |
| XM_021469325.1 | XP_021325000.1 | 4 - 5 | 2074 | 96 | ||
| GenBank mRNA | BC081474.1 | - | 2 - 3 | 157 | 96 | |
| BC093194.1 | - | 3 - 4 | 495 | 96 | ||
More Information
Set Membership: |
|

lrrc39
Genomic Map