Assay Details
| Gene Symbol: hmgb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017769.1 | NP_001017769.1 | 4 - 5 | 599 | 86 | |
| GenBank mRNA | BC093186.1 | - | 4 - 5 | 599 | 86 | |
| BC164996.1 | - | 3 - 4 | 499 | 86 | ||
More Information
Set Membership: |
|

hmgb3b
Genomic Map