Assay Details
| Gene Symbol: slc24a4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017770.1 | NP_001017770.1 | 9 - 10 | 957 | 100 | |
| XM_005158841.4 | XP_005158898.1 | 9 - 10 | 1293 | 100 | ||
| XM_009293194.3 | XP_009291469.1 | 10 - 11 | 1172 | 100 | ||
| XM_009293195.3 | XP_009291470.1 | 9 - 10 | 1293 | 100 | ||
| GenBank mRNA | BC093185.1 | - | 9 - 10 | 957 | 100 | |
More Information
Set Membership: |
|

slc24a4a
Genomic Map