Assay Details
| Gene Symbol: twist1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017820.1 | NP_001017820.1 | 1 - 2 | 745 | 110 | |
| GenBank mRNA | BC092796.1 | - | 1 - 2 | 745 | 110 | |
More Information
Set Membership: |
|

twist1b
Genomic Map