Assay Details
| Gene Symbol: grhpra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020525.1 | NP_001018361.1 | 5 - 6 | 607 | 102 | |
| GenBank mRNA | BC095040.1 | - | 5 - 6 | 607 | 102 | |
More Information
Set Membership: |
|

grhpra
Genomic Map