Assay Details
| Gene Symbol: grapa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020573.1 | NP_001018409.1 | 2 - 3 | 335 | 79 | |
| GenBank mRNA | BC095199.1 | - | 2 - 3 | 335 | 79 | |
| BC164308.1 | - | 2 - 3 | 214 | 79 | ||
More Information
Set Membership: |
|

grapa
Genomic Map