Assay Details
| Gene Symbol: dhrs7cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020657.1 | NP_001018493.1 | 3 - 4 | 354 | 93 | |
| XM_021480113.1 | XP_021335788.1 | 3 - 4 | 425 | 93 | ||
| GenBank mRNA | BC095682.1 | - | 3 - 4 | 354 | 93 | |
| BC153545.1 | - | 3 - 4 | 335 | 93 | ||
More Information
Set Membership: |
|

dhrs7cb
Genomic Map