Assay Details
| Gene Symbol: raver1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020667.2 | NP_001018503.2 | 2 - 3 | 415 | 92 | |
| NM_001309640.1 | NP_001296569.1 | 2 - 3 | 415 | 92 | ||
| XM_021470765.1 | XP_021326440.1 | 2 - 3 | 451 | 92 | ||
| GenBank mRNA | BC095698.1 | - | 2 - 3 | 415 | 92 | |
More Information
Set Membership: |
|

raver1
Genomic Map