Assay Details
| Gene Symbol: b3galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020687.1 | NP_001018523.1 | 8 - 9 | 1097 | 71 | |
| GenBank mRNA | BC095777.1 | - | 8 - 9 | 1097 | 71 | |
More Information
Set Membership: |
|

b3galnt2
Genomic Map