Assay Details
| Gene Symbol: lypd6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128329.1 | NP_001121801.1 | 4 - 5 | 456 | 133 | |
| XM_017357680.2 | XP_017213169.1 | 4 - 5 | 514 | 133 | ||
| XM_017357681.2 | XP_017213170.1 | 2 - 3 | 356 | 133 | ||
| XM_021478681.1 | XP_021334356.1 | 3 - 4 | 540 | 133 | ||
| XM_021478682.1 | XP_021334357.1 | 5 - 6 | 696 | 133 | ||
| XM_021478683.1 | XP_021334358.1 | 4 - 5 | 826 | 133 | ||
| XM_021478684.1 | XP_021334359.1 | 3 - 4 | 433 | 133 | ||
| GenBank mRNA | BC095889.1 | - | 4 - 5 | 444 | 133 | |
More Information
Set Membership: |
|

lypd6b
Genomic Map