Assay Details
| Gene Symbol: fahd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020728.1 | NP_001018564.1 | 1 | 755 | 124 | |
| GenBank mRNA | BC095057.1 | - | 1 | 755 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fahd1| Gene Symbol: fahd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020728.1 | NP_001018564.1 | 1 | 755 | 124 | |
| GenBank mRNA | BC095057.1 | - | 1 | 755 | 124 | |
Set Membership: |
|