Assay Details
| Gene Symbol: pop4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020749.2 | NP_001018585.1 | 6 - 7 | 612 | 97 | |
| XM_005166455.4 | XP_005166512.1 | 6 - 7 | 632 | 97 | ||
| GenBank mRNA | BC095328.1 | - | 6 - 7 | 613 | 97 | |
More Information
Set Membership: |
|

pop4
Genomic Map