Assay Details
| Gene Symbol: fpgt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020754.2 | NP_001018590.2 | 3 - 4 | 520 | 139 | |
| GenBank mRNA | BC095375.1 | - | 3 - 4 | 294 | 139 | |
| BC155808.1 | - | 3 - 4 | 520 | 139 | ||
More Information
Set Membership: |
|

fpgt
Genomic Map